Gene Transcripts

Gene Symbol
Trmt44
Entrez Gene ID
78890
Gene Name
tRNA methyltransferase 44
Gene Aliases
2310079F23Rik, Mettl19
Chromosome Location
Chr. 5: 35556208 - 35575070 on Build GRCm38
UniGene ID
Mm.98522
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030208.3 NP_084484.2 3 764
GenBank mRNA AK019101.1 - 3 755
AK046957.1 - 3 766
AK132402.1 - 3 765
BC152332.1 - 3 759 40141614

View Full Product Details