Gene Transcripts

Gene Symbol
Mageh1
Entrez Gene ID
75625
Gene Name
melanoma antigen, family H, 1
Gene Aliases
2010107K23Rik, APR-1, APR1
Chromosome Location
Chr. X: 153036166 - 153037563 on Build GRCm38
UniGene ID
Mm.6890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023788.3 NP_076277.1 1 358
GenBank mRNA AF319981.1 - 1 133
AK145866.1 - 1 320
BC051935.1 - 1 147 5714800
BC058380.1 - 1 137 6820349
BC060080.1 - 1 147 5714800
CD363769.1 - 1 358

View Full Product Details