Gene Transcripts

Gene Symbol
Prkrir
Entrez Gene ID
72981
Gene Name
protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor)
Gene Aliases
2900052B10Rik, Dap4, Rpkrir
Chromosome Location
Chr. 7: 98703230 - 98718062 on Build GRCm38
UniGene ID
Mm.4428
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028410.1 NP_082686.1 5 924
GenBank mRNA AK013663.1 - 5 1015
AK028175.1 - 1 30
BC049103.1 - 5 924 6400126

View Full Product Details