Gene Transcripts

Gene Symbol
ENOX2
Entrez Gene ID
10495
Gene Name
ecto-NOX disulfide-thiol exchanger 2
Gene Aliases
APK1, COVA1, tNOX
Chromosome Location
Chr. X: 130622330 - 130903317 on Build GRCh38
UniGene ID
Hs.171458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281736.1 NP_001268665.1 11 1749
NM_006375.3 NP_006366.2 12 1893
NM_182314.2 NP_872114.1 13 1982
XM_005262354.3 XP_005262411.1 12 1838
XM_011531244.2 XP_011529546.1 13 1984
XM_011531245.2 XP_011529547.1 13 1978
XM_011531247.2 XP_011529549.1 14 2110
XM_011531249.2 XP_011529551.1 11 1538
XM_011531251.2 XP_011529553.1 11 1643
XM_017029226.1 XP_016884715.1 15 2201
XM_017029227.1 XP_016884716.1 13 1935
XM_017029228.1 XP_016884717.1 12 1684
XM_017029229.1 XP_016884718.1 12 1887
GenBank mRNA AF207881.2 - 10 1503
AK000353.1 - 11 1666
AK289812.1 - 13 1874
AK289837.1 - 12 1785
BC019254.1 - 7 1018
BC139781.1 - 10 1566
BC140874.1 - 11 1663 9021742
S72904.1 - 5 659

View Full Product Details