Gene Transcripts

Gene Symbol
Lrriq4
Entrez Gene ID
68307
Gene Name
leucine-rich repeats and IQ motif containing 4
Gene Aliases
4930558O21Rik
Chromosome Location
Chr. 3: 30644506 - 30672431 on Build GRCm38
UniGene ID
Mm.389425
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290510.1 NP_001277439.1 5 1879
NM_026668.2 NP_080944.2 7 2042
GenBank mRNA AK015024.1 - 5 1830
AK015586.1 - 1 54
AK019775.1 - 7 2043
BC145806.1 - 7 1743 40131109

View Full Product Details