Gene Transcripts
- Gene Symbol
- Mfsd11
- Entrez Gene ID
- 69900
- Gene Name
- major facilitator superfamily domain containing 11
- Gene Aliases
- 2600014M03Rik
- Chromosome Location
- Chr. 11: 116852283 - 116875660 on Build GRCm38
- UniGene ID
- Mm.348004
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178620.3 |
NP_848735.1
|
2, 3 |
223 |
|
GenBank mRNA |
AF015191.1 |
- |
2, 3 |
189 |
|
AK031384.1 |
- |
2, 3 |
418 |
|
AK032043.1 |
- |
2, 3 |
419 |
|
AK080006.1 |
- |
2, 3 |
945 |
|
BC016209.1 |
- |
2, 3 |
207 |
|
BC079583.1 |
- |
2, 3 |
247 |
|
BC088988.1 |
- |
2, 3 |
223 |
6418757 |
BC100547.1 |
- |
3, 4 |
380 |
|
BC137970.1 |
- |
2, 3 |
210 |
8860989 |
BC137971.1 |
- |
2, 3 |
211 |
8860990 |
BC145068.1 |
- |
2, 3 |
212 |
|
View Full Product Details