Gene Transcripts

Gene Symbol
Nkapl
Entrez Gene ID
66707
Gene Name
NFKB activating protein-like
Gene Aliases
4921504I05Rik
Chromosome Location
Chr. 13: 21467047 - 21468501 on Build GRCm38
UniGene ID
Mm.116869
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025719.3 NP_079995.2 1 1097
GenBank mRNA AK014812.1 - 1 1084
AK160424.1 - 1 899
BC099550.1 - 1 1097 30918538

View Full Product Details