Gene Transcripts

Gene Symbol
Slc7a8
Entrez Gene ID
50934
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 8
Gene Aliases
AA408822, LAT2
Chromosome Location
Chr. 14: 54722215 - 54781886 on Build GRCm38
UniGene ID
Mm.276831
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016972.2 NP_058668.1 11 2835
GenBank mRNA AF171668.1 - 11 2490
BC059004.1 - 11 2835 6849059
Y19022.1 - 11 2503

View Full Product Details