Gene Transcripts

Gene Symbol
Mkrn1
Entrez Gene ID
54484
Gene Name
makorin, ring finger protein, 1
Gene Aliases
RFP
Chromosome Location
Chr. 6: 39397821 - 39420369 on Build GRCm38
UniGene ID
Mm.270484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018810.2 NP_061280.2 8 2119
GenBank mRNA AF192785.1 - 8 2144
AK029500.1 - 8 2159
AK077060.1 - 8 2120

View Full Product Details