Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019511.3 | NP_062384.1 | 3 | 1122 | |
GenBank mRNA | AF146524.1 | - | 3 | 1123 | |
AF209907.1 | - | 3 | 1082 | ||
AJ250491.1 | - | 3 | 1123 | ||
AK005513.1 | - | 4 | 1238 | ||
BC024765.1 | - | 3 | 1118 | 5364778 |