Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006762.2 | NP_006753.1 | 6 | 630 | |
XM_011542098.2 | XP_011540400.1 | 4 | 463 | ||
GenBank mRNA | AK292009.1 | - | 6 | 580 | |
AK297014.1 | - | 6 | 568 | ||
BC106896.1 | - | 6 | 589 | 40030918 | |
BC106897.1 | - | 6 | 589 | 40030919 | |
D42042.1 | - | 6 | 560 | ||
U30498.1 | - | 6 | 561 | ||
U51240.1 | - | 6 | 614 |