Gene Transcripts

Gene Symbol
Rgs7bp
Entrez Gene ID
52882
Gene Name
regulator of G-protein signalling 7 binding protein
Gene Aliases
A930030I01Rik, AI842293, D13Bwg1146e, R7bp
Chromosome Location
Chr. 13: 104945904 - 105054930 on Build GRCm38
UniGene ID
Mm.100348
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029879.2 NP_084155.2 6 3081
GenBank mRNA AK028323.1 - 6 2464
AK036240.1 - 6 2993
AK043529.1 - 3 1889

View Full Product Details