Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134095.2 | NP_598856.1 | 6 | 1117 | |
GenBank mRNA | AK005963.1 | - | 6 | 1073 | |
AK016347.1 | - | 6 | 1023 | ||
AK131703.1 | - | 6 | 1070 | ||
AK146155.1 | - | 6 | 1141 | ||
AK170731.1 | - | 7 | 1386 | ||
AK184624.1 | - | 1 | 296 | ||
BC022097.1 | - | 5 | 1072 | ||
BC056972.1 | - | 6 | 1117 | 6811343 | |
BC071205.1 | - | 6 | 1009 | 6515068 |