Gene Transcripts
- Gene Symbol
- Lair1
- Entrez Gene ID
- 52855
- Gene Name
- leukocyte-associated Ig-like receptor 1
- Gene Aliases
- 5133400O11Rik, BB115266, D7Bwg0421e, Lair-1
- Chromosome Location
- Chr. 7: 4007073 - 4073507 on Build GRCm38
- UniGene ID
- Mm.290880
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113474.1 |
NP_001106945.1
|
5, 6 |
670 |
|
NM_178611.6 |
NP_848726.1
|
4, 5 |
340 |
|
GenBank mRNA |
AF479685.1 |
- |
5, 6 |
605 |
|
AF491829.1 |
- |
5, 6 |
703 |
|
AF491830.1 |
- |
4, 5 |
252 |
|
AF491831.1 |
- |
5, 6 |
582 |
|
AK017222.1 |
- |
4, 5 |
344 |
|
AK033472.1 |
- |
4, 5 |
317 |
|
AK038574.1 |
- |
4, 5 |
303 |
|
AK043664.1 |
- |
4, 5 |
397 |
|
AK044465.1 |
- |
5, 6 |
641 |
|
AK050281.1 |
- |
5, 6 |
626 |
|
AK054158.1 |
- |
5, 6 |
749 |
|
AY392763.1 |
- |
5, 6 |
607 |
|
AY392764.1 |
- |
4, 5 |
277 |
|
AY392765.1 |
- |
3, 4 |
160 |
|
AY392766.1 |
- |
4, 5 |
490 |
|
BC025901.1 |
- |
5, 6 |
634 |
5253846 |
BY204263.1 |
- |
3, 4 |
284 |
|
View Full Product Details