Gene Transcripts

Gene Symbol
Sh3bp2
Entrez Gene ID
24055
Gene Name
SH3-domain binding protein 2
Gene Aliases
3BP2
Chromosome Location
Chr. 5: 34525784 - 34563639 on Build GRCm38
UniGene ID
Mm.5012
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145858.1 NP_001139330.1 13 2310
NM_001145859.1 NP_001139331.1 13 2313
NM_011893.3 NP_036023.3 13 2210
GenBank mRNA AK089455.1 - 13 2141
AK143548.1 - 13 2315
AK150265.1 - 13 2312
AK155103.1 - 13 2167
AK155898.1 - 13 2210
AK155970.1 - 13 2258
BC010198.1 - 13 2148 3967543
BC085497.1 - 13 2201 5696296
L14543.1 - 13 2144

View Full Product Details