Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013724.2 | NP_038752.2 | 13 | 2383 | |
GenBank mRNA | AB020741.1 | - | 13 | 2308 | |
AB035267.1 | - | 13 | 2061 | ||
BC068311.1 | - | 13 | 2358 | ||
BC139029.1 | - | 13 | 2150 | 8862051 | |
BC139031.1 | - | 13 | 2150 | 8862053 |