Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033314.3 | NP_201571.2 | 12 | 1771 | |
GenBank mRNA | AF323958.1 | - | 12 | 1640 | |
AK166724.1 | - | 12 | 1772 | ||
AK169706.1 | - | 12 | 1774 | ||
BC035200.1 | - | 1571 | 3158233 |