Gene Transcripts

Gene Symbol
Marcksl1
Entrez Gene ID
81520
Gene Name
MARCKS-like 1
Gene Aliases
F52, Mlp
Chromosome Location
Chr. 5: 147714163 - 147716486 on Build Rnor_6.0
UniGene ID
Rn.2486
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030862.2 NP_110489.1 2 960
GenBank mRNA AJ301677.1 - 2 853
BC081789.1 - 2 960 7134056

Gene Symbol
LOC102550530
Entrez Gene ID
102550530
Gene Name
MARCKS-related protein-like
Gene Aliases
Chromosome Location
Chr. 1: 84792063 - 84794912 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759283.1 XP_008757505.1 2 919

Gene Symbol
LOC103689966
Entrez Gene ID
103689966
Gene Name
MARCKS-related protein-like
Gene Aliases
Chromosome Location
Chr. 1: 84811226 - 84812702 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759211.1 XP_008757433.1 1 925

View Full Product Details