Gene Transcripts

Gene Symbol
Prdx1
Entrez Gene ID
117254
Gene Name
peroxiredoxin 1
Gene Aliases
Hbp23
Chromosome Location
Chr. 5: 135536413 - 135551986 on Build Rnor_6.0
UniGene ID
Rn.2845
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057114.1 NP_476455.1 4 390
GenBank mRNA BC058450.1 - 4 410
BC088118.1 - 4 388 7324630
D30035.1 - 4 390
FQ219859.1 - 4 407

View Full Product Details