Gene Transcripts

Gene Symbol
Blcap
Entrez Gene ID
171113
Gene Name
bladder cancer associated protein
Gene Aliases
Chromosome Location
Chr. 3: 154040165 - 154042096 on Build Rnor_6.0
UniGene ID
Rn.127812
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133582.3 NP_598266.1 1 380
GenBank mRNA AB071598.1 - 1 348
BC087661.1 - 1 173 7313852

View Full Product Details