Gene Transcripts
- Gene Symbol
- Clmp
- Entrez Gene ID
- 286939
- Gene Name
- CXADR-like membrane protein
- Gene Aliases
- ACAM, Asam, Ol16
- Chromosome Location
- Chr. 8: 44847157 - 44975460 on Build Rnor_6.0
- UniGene ID
- Rn.163013
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173154.1 |
NP_775177.1
|
2, 3 |
277 |
|
GenBank mRNA |
AF302047.1 |
- |
2, 3 |
277 |
|
View Full Product Details