Gene Transcripts

Gene Symbol
LST1
Entrez Gene ID
7940
Gene Name
leukocyte specific transcript 1
Gene Aliases
B144, D6S49E, LST-1
Chromosome Location
Chr. 6: 31586179 - 31588909 on Build GRCh38
UniGene ID
Hs.436066
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166538.1 NP_001160010.1 3 279
NM_007161.3 NP_009092.3 4 372
NM_205837.2 NP_995309.2 4 458
NM_205838.2 NP_995310.2 4 395
NM_205839.2 NP_995311.2 5 488
NM_205840.2 NP_995312.2 3 328
NR_029461.1 3 372
NR_029462.1 3 327
XM_006715206.3 XP_006715269.1 5 501
XM_006715209.3 XP_006715272.1 4 404
XM_006715210.3 XP_006715273.1 4 459
XM_011514914.2 XP_011513216.1 5 482
GenBank mRNA AF000424.1 - 4 458
AF000425.1 - 5 466
AF000426.1 - 3 327
AJ705986.1 - 3 309
AV711621.1 - 3 291
AV715235.1 - 4 450
AY852265.1 - 3 183
AY852266.1 - 4 272
BC103855.1 - 3 263 40003000
BC103856.1 - 3 263 40003004
BI838080.1 - 4 308
BX647293.1 - 2 1554
DQ099382.1 - 3 230
Y18486.1 - 3 280
Y18487.1 - 3 328
Y18488.1 - 3 213
Y18489.1 - 2 256
Y18490.1 - 2 190

View Full Product Details