Gene Transcripts

Gene Symbol
Nrn1
Entrez Gene ID
83834
Gene Name
neuritin 1
Gene Aliases
Nrn
Chromosome Location
Chr. 17: 27925054 - 28002946 on Build Rnor_6.0
UniGene ID
Rn.3546
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053346.1 NP_445798.1 2 599
GenBank mRNA BC087582.1 - 2 536
FQ213150.1 - 1 137
U88958.1 - 2 599

View Full Product Details