Gene Transcripts

Gene Symbol
Pik3c2g
Entrez Gene ID
116720
Gene Name
phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 gamma
Gene Aliases
Chromosome Location
Chr. 4: 173732196 - 174102502 on Build Rnor_6.0
UniGene ID
Rn.14870
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053923.1 NP_446375.1 2 425
XM_008763348.1 XP_008761570.1 2 477
GenBank mRNA AB009636.1 - 2 425

View Full Product Details