Gene Transcripts

Gene Symbol
Rbbp9
Entrez Gene ID
29459
Gene Name
retinoblastoma binding protein 9
Gene Aliases
Chromosome Location
Chr. 3: 138701579 - 138708332 on Build Rnor_6.0
UniGene ID
Rn.162693
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019219.2 NP_062092.2 5 1441
GenBank mRNA AF025819.2 - 5 1441

View Full Product Details