Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020100.2 | NP_064485.2 | 3 | 721 | |
GenBank mRNA | AB042889.1 | - | 3 | 719 | |
AF181552.2 | - | 3 | 721 | ||
FQ214551.1 | - | 3 | 715 | ||
FQ220330.1 | - | 3 | 702 | ||
FQ220789.1 | - | 3 | 723 | ||
FQ229563.1 | - | 3 | 740 |