Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173148.1 | NP_775171.1 | 377 | ||
XM_008761579.1 | XP_008759801.1 | 5 | 482 | ||
GenBank mRNA | M76734.1 | - | 377 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008761619.1 | XP_008759841.1 | 383 |