Gene Transcripts

Gene Symbol
CLEC5A
Entrez Gene ID
23601
Gene Name
C-type lectin domain family 5 member A
Gene Aliases
CLECSF5, MDL-1, MDL1
Chromosome Location
Chr. 7: 141927356 - 141947009 on Build GRCh38
UniGene ID
Hs.446235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013252.2 NP_037384.1 3, 4 330
XM_017011916.1 XP_016867405.1 3, 4 356
GenBank mRNA AF139768.1 - 3, 4 284
AJ271684.1 - 3, 4 330
AK292766.1 - 3, 4 319
AK313703.1 - 3, 4 319
BC093714.1 - 2, 3 154 7939559
BC112099.1 - 2, 3 154 8327567
BC113098.1 - 3, 4 272 40076723

View Full Product Details