Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013262.3 | NP_037394.2 | 2 | 340 | |
XM_005249032.2 | XP_005249089.1 | 2 | 340 | ||
XM_017010789.1 | XP_016866278.1 | 2 | 340 | ||
GenBank mRNA | AF006003.1 | - | 2 | 253 | |
AF187016.2 | - | 2 | 277 | ||
AF258586.1 | - | 2 | 317 | ||
AK074391.1 | - | 2 | 340 | ||
AK314648.1 | - | 2 | 340 | ||
BC002860.2 | - | 2 | 321 | 3638617 | |
BT007055.1 | - | 2 | 103 |