Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300795.1 | NP_001287724.1 | 8 | 1743 | |
NM_003340.6 | NP_003331.1 | 8 | 2050 | ||
NM_181886.3 | NP_871615.1 | 8 | 2258 | ||
NM_181887.2 | NP_871616.1 | 8 | 2028 | ||
NM_181888.3 | NP_871617.1 | 8 | 1907 | ||
NM_181889.2 | NP_871618.1 | 8 | 1930 | ||
NM_181890.2 | NP_871619.1 | 8 | 1872 | ||
NM_181891.2 | NP_871620.1 | 8 | 2215 | ||
NM_181892.3 | NP_871621.1 | 8 | 2290 | ||
NM_181893.2 | NP_871622.1 | 7 | 1720 | ||
XM_017008584.1 | XP_016864073.1 | 7 | 2082 | ||
GenBank mRNA | AK095822.1 | - | 8 | 2205 | |
AK127304.1 | - | 6 | 4663 | ||
AL050186.1 | - | 1 | 593 | ||
BC037894.2 | - | 8 | 2028 | 5275356 |