Gene Transcripts

Gene Symbol
RBMXL2
Entrez Gene ID
27288
Gene Name
RNA binding motif protein, X-linked like 2
Gene Aliases
HNRNPG-T, HNRNPGT, HNRPGT
Chromosome Location
Chr. 11: 7088934 - 7091148 on Build GRCh38
UniGene ID
Hs.121605
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014469.4 NP_055284.3 1 1467
GenBank mRNA AF069682.2 - 1 1373
AK292271.1 - 1 1374
BC057796.1 - 1 1298 5261317

View Full Product Details