Gene Transcripts

Gene Symbol
OR10AD1
Entrez Gene ID
121275
Gene Name
olfactory receptor family 10 subfamily AD member 1
Gene Aliases
OR10AD1P, OR12-1
Chromosome Location
Chr. 12: 48202339 - 48203292 on Build GRCh38
UniGene ID
Hs.446790
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004134.1 NP_001004134.1 1 421
GenBank mRNA BC136757.1 - 1 516
BC136758.1 - 1 516 9020748

View Full Product Details