Gene Transcripts

Gene Symbol
NAV3
Entrez Gene ID
89795
Gene Name
neuron navigator 3
Gene Aliases
POMFIL1, STEERIN3, unc53H3
Chromosome Location
Chr. 12: 77571862 - 78213012 on Build GRCh38
UniGene ID
Hs.655301
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024383.1 NP_001019554.1 34 6325
NM_014903.5 NP_055718.4 33 6259
XM_005269215.3 XP_005269272.1 32 10239
XM_011538940.2 XP_011537242.1 32 10219
XM_011538941.2 XP_011537243.1 31 10197
XM_011538944.2 XP_011537246.1 31 9727
XM_017020164.1 XP_016875653.1 33 10307
XM_017020165.1 XP_016875654.1 32 10265
XM_017020166.1 XP_016875655.1 34 6314
XM_017020167.1 XP_016875656.1 33 6245
XM_017020168.1 XP_016875657.1 33 9838
XM_017020169.1 XP_016875658.1 32 9795
XM_017020170.1 XP_016875659.1 30 9705
XM_017020171.1 XP_016875660.1 26 9621
XM_017020172.1 XP_016875661.1 23 5130
XM_017020173.1 XP_016875662.1 25 9130
XM_017020174.1 XP_016875663.1 24 9088
XM_017020175.1 XP_016875664.1 24 4943
GenBank mRNA AB023155.2 - 18 2769
AF397731.1 - 32 5843
AJ488201.1 - 34 6197
AK298525.1 - 16 1952

View Full Product Details