Gene Transcripts

Gene Symbol
Mrps10
Entrez Gene ID
363187
Gene Name
mitochondrial ribosomal protein S10
Gene Aliases
Ac1179
Chromosome Location
Chr. 9: 15637427 - 15651154 on Build Rnor_6.0
UniGene ID
Rn.101558
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008859.1 NP_001008859.1 2, 3 93
GenBank mRNA AY310148.1 - 2, 3 93

View Full Product Details