Gene Transcripts

Gene Symbol
Vom1r22
Entrez Gene ID
494252
Gene Name
vomeronasal 1 receptor 22
Gene Aliases
V1rf2
Chromosome Location
Chr. 1: 61625441 - 61626454 on Build Rnor_6.0
UniGene ID
Rn.129446
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008900.1 NP_001008900.1 1 453
GenBank mRNA AY510355.1 - 1 453

View Full Product Details