Gene Transcripts

Gene Symbol
Mrgprb13
Entrez Gene ID
404643
Gene Name
MAS-related GPR, member B13
Gene Aliases
MrgB8, Mrgprb8
Chromosome Location
Chr. 1: 103630249 - 103631235 on Build Rnor_6.0
UniGene ID
Rn.141967
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002283.1 NP_001002283.1 501

Gene Symbol
LOC100361204
Entrez Gene ID
100361204
Gene Name
mas-related G-protein coupled receptor member B8-like
Gene Aliases
Chromosome Location
Chr. 1: 103621813 - 103622816 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759464.1 XP_008757686.1 1 518

Gene Symbol
LOC100912474
Entrez Gene ID
100912474
Gene Name
mas-related G-protein coupled receptor member B8-like
Gene Aliases
Chromosome Location
Chr. 1: 103681845 - 103682937 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748862.2 XP_003748910.1 1 518

View Full Product Details