Gene Transcripts

Gene Symbol
MLEC
Entrez Gene ID
9761
Gene Name
malectin
Gene Aliases
KIAA0152
Chromosome Location
Chr. 12: 120687124 - 120701864 on Build GRCh38
UniGene ID
Hs.744910
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303627.1 NP_001290556.1 5 5417
NM_001303628.1 NP_001290557.1 3 5465
NM_014730.3 NP_055545.1 5 5700
XM_011539031.1 XP_011537333.1 5 10115
XM_011539032.1 XP_011537334.1 6 9803
GenBank mRNA BE622248.1 - 1 315
CR936818.1 - 2 4870
D63486.1 - 5 5657

View Full Product Details