Gene Transcripts

Gene Symbol
OR2A2
Entrez Gene ID
442361
Gene Name
olfactory receptor family 2 subfamily A member 2
Gene Aliases
OR2A17P, OR2A2P, OR7-11, OST008
Chromosome Location
Chr. 7: 144109583 - 144110539 on Build GRCh38
UniGene ID
Hs.553841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005480.2 NP_001005480.2 1 931
GenBank mRNA BC136742.1 - 1 1000

View Full Product Details