Gene Transcripts

Gene Symbol
Lrrn2
Entrez Gene ID
16980
Gene Name
leucine rich repeat protein 2, neuronal
Gene Aliases
5730406J09Rik, NLRR-2
Chromosome Location
Chr. 1: 132880355 - 132940005 on Build GRCm38
UniGene ID
Mm.39640
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010732.4 NP_034862.1 2 3167
GenBank mRNA AK019955.1 - 1 139
AK036271.1 - 2 3169
AK039216.1 - 2 3168
AY362823.1 - 1 2756
BB342744.1 - 1 92
BC056458.1 - 2 2909 5685881

View Full Product Details