Gene Transcripts
- Gene Symbol
- NRXN2
- Entrez Gene ID
- 9379
- Gene Name
- neurexin 2
- Gene Aliases
- Chromosome Location
- Chr. 11: 64606174 - 64723188 on Build GRCh38
- UniGene ID
- Hs.372938
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015080.3 |
NP_055895.1
|
22, 23 |
4725 |
|
NM_138732.2 |
NP_620060.1
|
19, 20 |
4515 |
|
NM_138734.2 |
NP_620063.1
|
6, 7 |
1637 |
|
XM_005274400.3 |
XP_005274457.1
|
21, 22 |
4257 |
|
XM_011545370.1 |
XP_011543672.1
|
21, 22 |
4287 |
|
XM_011545371.1 |
XP_011543673.1
|
21, 22 |
4284 |
|
XM_011545373.1 |
XP_011543675.1
|
21, 22 |
4266 |
|
XM_011545375.1 |
XP_011543677.1
|
20, 21 |
4242 |
|
XM_011545385.1 |
XP_011543687.1
|
14, 15 |
2632 |
|
XM_017018563.1 |
XP_016874052.1
|
20, 21 |
4257 |
|
XM_017018564.1 |
XP_016874053.1
|
20, 21 |
4254 |
|
XM_017018565.1 |
XP_016874054.1
|
20, 21 |
4251 |
|
XM_017018566.1 |
XP_016874055.1
|
20, 21 |
4215 |
|
XM_017018567.1 |
XP_016874056.1
|
20, 21 |
4197 |
|
XM_017018568.1 |
XP_016874057.1
|
20, 21 |
4188 |
|
XM_017018569.1 |
XP_016874058.1
|
19, 20 |
4185 |
|
XM_017018572.1 |
XP_016874061.1
|
21, 22 |
3582 |
|
XM_017018574.1 |
XP_016874063.1
|
20, 21 |
3552 |
|
XM_017018575.1 |
XP_016874064.1
|
20, 21 |
3549 |
|
XM_017018576.1 |
XP_016874065.1
|
16, 17 |
3197 |
|
XM_017018577.1 |
XP_016874066.1
|
14, 15 |
2623 |
|
GenBank mRNA |
AB023138.2 |
- |
20, 21 |
4492 |
|
AB035266.1 |
- |
21, 22 |
4377 |
|
AB177862.1 |
- |
18, 19 |
3429 |
|
AK098211.1 |
- |
2, 3 |
1334 |
|
BC150275.1 |
- |
20, 21 |
4492 |
9007249 |
View Full Product Details