Gene Transcripts
- Gene Symbol
- TRIM2
- Entrez Gene ID
- 23321
- Gene Name
- tripartite motif containing 2
- Gene Aliases
- CMT2R, RNF86
- Chromosome Location
- Chr. 4: 153152281 - 153339322 on Build GRCh38
- UniGene ID
- Hs.435711
- Species
- Homo sapiens
View Full Product Details