Gene Transcripts

Gene Symbol
UPF2
Entrez Gene ID
26019
Gene Name
UPF2 regulator of nonsense transcripts homolog (yeast)
Gene Aliases
HUPF2, RENT2, smg-3
Chromosome Location
Chr. 10: 11920022 - 12043170 on Build GRCh38
UniGene ID
Hs.370689
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015542.3 NP_056357.1 2 314
NM_080599.2 NP_542166.1 2 484
XM_011519449.2 XP_011517751.1 2 362
GenBank mRNA AB037829.1 - 1 683
AF301013.1 - 2 307
AF318574.1 - 2 284
AK310925.1 - 2 273
AY013249.1 - 1 233
BC090919.1 - 2 282
BC114964.1 - 2 278 40082739
BC115737.1 - 2 278 40082728
DA188353.1 - 2 314

View Full Product Details