Gene Transcripts

Gene Symbol
RIBC2
Entrez Gene ID
26150
Gene Name
RIB43A domain with coiled-coils 2
Gene Aliases
C22orf11, TRIB
Chromosome Location
Chr. 22: 45413691 - 45432422 on Build GRCh38
UniGene ID
Hs.475110
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015653.4 NP_056468.3 3 671
XM_005261524.4 XP_005261581.1 3 496
XM_017028766.1 XP_016884255.1 3 593
GenBank mRNA AK098586.1 - 3 671
AK225722.1 - 3 652
AL442116.1 - 3 622
BC003024.1 - 3 633 2822570
BC009904.2 - 3 633 2822570
CR456438.1 - 1 265
CR457398.1 - 1 256

View Full Product Details