Gene Transcripts
- Gene Symbol
- PDGFC
- Entrez Gene ID
- 56034
- Gene Name
- platelet derived growth factor C
- Gene Aliases
- FALLOTEIN, SCDGF
- Chromosome Location
- Chr. 4: 156761611 - 156971394 on Build GRCh38
- UniGene ID
- Hs.570855
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016205.2 |
NP_057289.1
|
1, 2 |
607 |
|
XM_017008456.1 |
XP_016863945.1
|
1, 2 |
569 |
|
GenBank mRNA |
AB033831.1 |
- |
1, 2 |
442 |
|
AF091434.1 |
- |
1, 2 |
607 |
|
AF244813.1 |
- |
1, 2 |
304 |
|
AF260738.1 |
- |
1, 2 |
278 |
|
AK313817.1 |
- |
1, 2 |
337 |
|
AY358493.1 |
- |
1, 2 |
400 |
|
BC094746.1 |
- |
1, 2 |
208 |
|
BC136662.1 |
- |
1, 2 |
528 |
9020652 |
View Full Product Details