Gene Transcripts

Gene Symbol
Trip13
Entrez Gene ID
292206
Gene Name
thyroid hormone receptor interactor 13
Gene Aliases
Chromosome Location
Chr. 1: 31967980 - 32012923 on Build Rnor_6.0
UniGene ID
Rn.11751
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011930.1 NP_001011930.1 8 830
XM_008758681.1 XP_008756903.1 8 832
GenBank mRNA BC083900.1 - 8 830

View Full Product Details