Gene Transcripts

Gene Symbol
Pdrg1
Entrez Gene ID
296278
Gene Name
p53 and DNA damage regulated 1
Gene Aliases
RGD1305096
Chromosome Location
Chr. 3: 148487056 - 148493225 on Build Rnor_6.0
UniGene ID
Rn.101752
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014762.1 NP_001014762.1 5 1178
GenBank mRNA BC082021.1 - 5 1178

View Full Product Details