Gene Transcripts

Gene Symbol
Wscd1
Entrez Gene ID
287466
Gene Name
WSC domain containing 1
Gene Aliases
RGD1308212
Chromosome Location
Chr. 10: 58342393 - 58371199 on Build Rnor_6.0
UniGene ID
Rn.198297
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024234.1 NP_001019405.1 6 1543
GenBank mRNA BC094520.1 - 6 1543

View Full Product Details