Gene Transcripts

Gene Symbol
Olr67
Entrez Gene ID
365324
Gene Name
olfactory receptor 67
Gene Aliases
Chromosome Location
Chr. 1: 168117526 - 168118464 on Build Rnor_6.0
UniGene ID
Rn.141916
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001279.1 NP_001001279.1 1 702

View Full Product Details