Gene Transcripts

Gene Symbol
STH
Entrez Gene ID
246744
Gene Name
saitohin
Gene Aliases
MAPTIT
Chromosome Location
Chr. 17: 45999250 - 45999694 on Build GRCh38
UniGene ID
Hs.661831
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007532.2 NP_001007533.1 1 149
GenBank mRNA AY179170.1 - 1 119
BC130319.1 - 1 149 40146350
BC130321.1 - 1 149 40146352

View Full Product Details