Gene Transcripts

Gene Symbol
Mettl7b
Entrez Gene ID
366792
Gene Name
methyltransferase like 7B
Gene Aliases
RGD1305205
Chromosome Location
Chr. 7: 3383876 - 3386522 on Build Rnor_6.0
UniGene ID
Rn.145341
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024276.1 NP_001019447.1 1 408
GenBank mRNA BC092587.1 - 1 401
BC092590.1 - 1 408 7371843

View Full Product Details